ENA245485452 Enamine

N-ethyl-3,5-dimethyl-N-(2-methylpropyl)-1H-pyrazole-4-sulfonamide hydrochloride

  • MDL number MFCD12913148
  • Cas Number : 1239484-85-5
  • Molecular Weight : 295.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52700
Availability details for
Quantity
  • Availability
  • Add to Cart