ENA245485455 Enamine

2-chloro-N-cyclopropyl-N-[(thiophen-3-yl)methyl]acetamide

  • MDL number MFCD12913149
  • Cas Number : 1197909-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-52703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52703
Availability details for
Quantity
  • Availability
  • Add to Cart