ENA245485460 Enamine

4-bromo-1H-pyrrolo[2,3-b]pyridine-3-sulfonyl chloride

  • MDL number MFCD12913150
  • Cas Number : 1210227-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52708
Availability details for
Quantity
  • Availability
  • Add to Cart