ENA245485461 Enamine

N-methyl-2,3-dihydro-1H-inden-2-amine

  • MDL number MFCD12027038
  • Cas Number : 24445-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52709

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52709
Availability details for
Quantity
  • Availability
  • Add to Cart