ENA245485464 Enamine

4-chloro-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00129178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 261-263
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52720

Safety Information

signalword Danger
pcodes P264,P270,P280,P301+P310,P305+P351+P338,P330,P337+P313,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-52720
pictograms GHS06
hstatements H301,H319
Availability details for
Quantity
  • Availability
  • Add to Cart