ENA245485471 Enamine

1-(2-methoxy-2-oxoethyl)-2,5-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09731734
  • Cas Number : 953737-58-1
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52729
Availability details for
Quantity
  • Availability
  • Add to Cart