ENA245485475 Enamine

5-(2-chloroacetyl)thiophene-3-carboxamide

  • MDL number MFCD12913155
  • Cas Number : 1197469-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-52734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52734
Availability details for
Quantity
  • Availability
  • Add to Cart