ENA245485479 Enamine

4-bromo-2-chloro-N-methylaniline

  • MDL number MFCD09745401
  • Cas Number : 944131-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52739
Availability details for
Quantity
  • Availability
  • Add to Cart