ENA245485488 Enamine

1-(2,4-difluorophenoxy)propan-2-amine

  • MDL number MFCD09735791
  • Cas Number : 953751-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52751

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-52751
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart