ENA245485491 Enamine

6-methyl-1H,4H,7H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD13195854
  • Cas Number : 30129-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52755
Availability details for
Quantity
  • Availability
  • Add to Cart