ENA245485495 Enamine

3-[(1-methyl-1H-imidazol-2-yl)sulfanyl]propanoic acid

  • MDL number MFCD09738265
  • Cas Number : 919467-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52761
Availability details for
Quantity
  • Availability
  • Add to Cart