ENA245485500 Enamine

2-(propane-1-sulfonyl)propanoic acid

  • MDL number MFCD09940594
  • Cas Number : 854678-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52768
Availability details for
Quantity
  • Availability
  • Add to Cart