ENA245485505 Enamine

1-(3,4-dichlorophenyl)-2-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD09803058
  • Cas Number : 10006-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52789
Availability details for
Quantity
  • Availability
  • Add to Cart