ENA245485519 Enamine

1-phenyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD11646040
  • Cas Number : 1152979-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52812
Availability details for
Quantity
  • Availability
  • Add to Cart