ENA245485526 Enamine

1-benzyl-4-phenylpiperazine

  • MDL number MFCD00811058
  • Cas Number : 3074-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52824
Availability details for
Quantity
  • Availability
  • Add to Cart