ENA245485528 Enamine

1-{[(prop-2-yn-1-yl)carbamoyl]amino}-2,3-dihydro-1H-indene-1-carboxylic acid

  • MDL number MFCD12913168
  • Cas Number : 1197578-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52826
Availability details for
Quantity
  • Availability
  • Add to Cart