ENA245485529 Enamine

1-(5-methyl-1,2-oxazol-3-yl)-3-(prop-2-yn-1-yl)urea

  • MDL number MFCD12913169
  • Cas Number : 1197564-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52827
Availability details for
Quantity
  • Availability
  • Add to Cart