ENA245485535 Enamine

5-methyl-1-(3-nitrophenyl)-2,3-dihydro-1H-imidazol-2-one

  • MDL number MFCD12913173
  • Cas Number : 13870-56-9
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52835
Availability details for
Quantity
  • Availability
  • Add to Cart