ENA245485537 Enamine

N-hydroxy-2-(4-methylphenyl)acetamide

  • MDL number MFCD11204735
  • Cas Number : 2779-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155...157
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52837

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-52837
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart