ENA245485541 Enamine

4H,5H-[1,2,4]triazolo[1,5-a]pyrimidin-5-one

  • MDL number MFCD12913174
  • Cas Number : 14384-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52841
Availability details for
Quantity
  • Availability
  • Add to Cart