ENA245485542 Enamine

4-fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD08544171
  • Cas Number : 936220-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 286...288
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52842

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52842
Availability details for
Quantity
  • Availability
  • Add to Cart