ENA245485543 Enamine

2-(2-amino-5-methylphenoxy)-N,N-dimethylacetamide

  • MDL number MFCD09945239
  • Cas Number : 1016824-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52843
Availability details for
Quantity
  • Availability
  • Add to Cart