ENA245485547 Enamine

1-benzyl-2,3-dihydro-1H-indol-5-amine hydrochloride

  • MDL number MFCD12913177
  • Cas Number : 1210277-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52865
Availability details for
Quantity
  • Availability
  • Add to Cart