ENA245485549 Enamine

2-{4-[(1-methyl-1H-imidazol-2-yl)methoxy]phenyl}ethan-1-amine

  • MDL number MFCD11892393
  • Cas Number : 1181346-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52868
Availability details for
Quantity
  • Availability
  • Add to Cart