ENA245485553 Enamine

4-bromo-2-(4-fluorophenoxy)benzoic acid

  • MDL number MFCD11551615
  • Cas Number : 1099687-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-52872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52872
Availability details for
Quantity
  • Availability
  • Add to Cart