ENA245485554 Enamine

2,3-dioxo-2,3-dihydro-1H-indole-5-carboxylic acid

  • MDL number MFCD00967204
  • Cas Number : 25128-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 292-295
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52873
Availability details for
Quantity
  • Availability
  • Add to Cart