ENA245485562 Enamine

2,2,2-trifluoroethyl N-(6-methoxypyridin-3-yl)carbamate

  • MDL number MFCD11099789
  • Cas Number : 1087797-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52884
Availability details for
Quantity
  • Availability
  • Add to Cart