ENA245485565 Enamine

2,2,2-trifluoroethyl N-(1-ethyl-1H-pyrazol-4-yl)carbamate

  • MDL number MFCD12134736
  • Cas Number : 1156304-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52888
Availability details for
Quantity
  • Availability
  • Add to Cart