ENA245485566 Enamine

2,2,2-trifluoroethyl N-[1-(2-methoxyethyl)-1H-pyrazol-4-yl]carbamate

  • MDL number MFCD13195860
  • Cas Number : 1211189-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52890
Availability details for
Quantity
  • Availability
  • Add to Cart