ENA245485573 Enamine

3-amino-3-(3-chlorophenyl)propanoic acid

  • MDL number MFCD00463160
  • Cas Number : 68208-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227...229
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52901

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-52901
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart