ENA245485586 Enamine

[2-(2-methyl-1H-imidazol-1-yl)pyridin-3-yl]methanamine

  • MDL number MFCD09742922
  • Cas Number : 954569-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52916

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52916
Availability details for
Quantity
  • Availability
  • Add to Cart