ENA245485613 Enamine

2-(3-methoxyphenyl)propanenitrile

  • MDL number MFCD11036520
  • Cas Number : 25310-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52967
Availability details for
Quantity
  • Availability
  • Add to Cart