ENA245485615 Enamine

[(4-bromophenyl)methyl](pyridin-2-ylmethyl)amine

  • MDL number MFCD05740280
  • Cas Number : 774556-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52969

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52969
Availability details for
Quantity
  • Availability
  • Add to Cart