ENA245485622 Enamine

N-(4-amino-2-methylphenyl)-2-iodobenzamide

  • MDL number MFCD09044711
  • Cas Number : 926221-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-52984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52984
Availability details for
Quantity
  • Availability
  • Add to Cart