ENA245485626 Enamine

1-(3-amino-4-fluorophenyl)pyrrolidine-2,5-dione

  • MDL number MFCD09735900
  • Cas Number : 953741-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52988
Availability details for
Quantity
  • Availability
  • Add to Cart