ENA245485634 Enamine

2-amino-5-chloro-N-(2-methylpropyl)benzamide

  • MDL number MFCD09050269
  • Cas Number : 34810-93-0
  • Molecular Weight : 226.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53010
Availability details for
Quantity
  • Availability
  • Add to Cart