ENA245485651 Enamine

(2S)-2-({thieno[2,3-d]pyrimidin-4-yl}amino)propanoic acid

  • MDL number MFCD12913199
  • Cas Number : 63895-63-6
  • Molecular Weight : 223.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53033
Availability details for
Quantity
  • Availability
  • Add to Cart