ENA245485657 Enamine

2-methyl-2,3-dihydro-1H-isoindol-5-amine

  • MDL number MFCD09263790
  • Cas Number : 158944-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61...63
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53042

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53042
Availability details for
Quantity
  • Availability
  • Add to Cart