ENA245485663 Enamine

2-(3-methoxyphenyl)propanoic acid

  • MDL number MFCD11036874
  • Cas Number : 3146-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53048

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-53048
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart