ENA245485675 Enamine

(1R)-1-(3,4-dimethoxyphenyl)ethan-1-ol

  • MDL number MFCD09863680
  • Cas Number : 120466-67-3
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53064
Availability details for
Quantity
  • Availability
  • Add to Cart