ENA245485681 Enamine

(1S)-1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)ethan-1-amine

  • MDL number MFCD09863770
  • Cas Number : 1212386-50-9
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53071
Availability details for
Quantity
  • Availability
  • Add to Cart