ENA245485693 Enamine

(1S)-1-(4-methanesulfonylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD12913202
  • Cas Number : 1212202-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53083
Availability details for
Quantity
  • Availability
  • Add to Cart