ENA245485699 Enamine

5-propylimidazolidine-2,4-dione

  • MDL number MFCD00059721
  • Cas Number : 18227-41-3
  • Molecular Weight : 142.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-138
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21324
Availability details for
Quantity
  • Availability
  • Add to Cart