ENA245485704 Enamine

6-bromo-2-[2-(4-methoxyphenyl)ethenyl]quinoline-4-carboxylic acid

  • Cas Number : 926222-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53100
Availability details for
Quantity
  • Availability
  • Add to Cart