ENA245485711 Enamine

1-(5-amino-2-methoxyphenyl)-3-(2-methoxyphenyl)urea

  • MDL number MFCD09045546
  • Cas Number : 926254-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53107
Availability details for
Quantity
  • Availability
  • Add to Cart