ENA245485715 Enamine

3-(1-benzofuran-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD10693652
  • Cas Number : 1152541-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-53111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53111
Availability details for
Quantity
  • Availability
  • Add to Cart