ENA245485721 Enamine

[2-(3-bromophenoxy)ethyl]dimethylamine

  • MDL number MFCD03993547
  • Cas Number : 221915-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53122
Availability details for
Quantity
  • Availability
  • Add to Cart