ENA245485727 Enamine

3-(4-aminophenyl)-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD12913206
  • Cas Number : 1197965-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53128
Availability details for
Quantity
  • Availability
  • Add to Cart