ENA245485730 Enamine

propane-2-sulfonyl fluoride

  • MDL number MFCD12913207
  • Cas Number : 63805-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53133

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53133
Availability details for
Quantity
  • Availability
  • Add to Cart