ENA245485733 Enamine

tert-butyl 3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate

  • MDL number MFCD13568915
  • Cas Number : 478837-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53136

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53136
Availability details for
Quantity
  • Availability
  • Add to Cart